Gene Transcripts

Gene Symbol
Nme3
Entrez Gene ID
85269
Gene Name
NME/NM23 nucleoside diphosphate kinase 3
Gene Aliases
NM23-dr
Chromosome Location
Chr. 10: 14258232 - 14259365 on Build Rnor_6.0
UniGene ID
Rn.8785
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053507.1 NP_445959.1 3 245
XM_008767598.1 XP_008765820.1 3 357
GenBank mRNA AY017337.1 - 3 245

View Full Product Details