Gene Transcripts

Gene Symbol
P3h1
Entrez Gene ID
114200
Gene Name
prolyl 3-hydroxylase 1
Gene Aliases
Gros1, Lepre1
Chromosome Location
Chr. 5: 138279597 - 138294280 on Build Rnor_6.0
UniGene ID
Rn.13741
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053667.1 NP_446119.1 5 955
GenBank mRNA AF087433.1 - 5 955
BC161838.1 - 5 989 9101602

View Full Product Details