Gene Transcripts

Gene Symbol
Atxn10
Entrez Gene ID
170821
Gene Name
ataxin 10
Gene Aliases
Sca10
Chromosome Location
Chr. 7: 126228416 - 126351728 on Build Rnor_6.0
UniGene ID
Rn.6524
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133313.2 NP_579847.1 2 226
GenBank mRNA AJ301634.1 - 2 335
BC062087.1 - 2 226 5621521

View Full Product Details