Gene Transcripts

Gene Symbol
Fut11
Entrez Gene ID
286971
Gene Name
fucosyltransferase 11 (alpha (1,3) fucosyltransferase)
Gene Aliases
Chromosome Location
Chr. 15: 4031504 - 4035064 on Build Rnor_6.0
UniGene ID
Rn.24040
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173308.2 NP_775430.2 2 1276
GenBank mRNA AJ535753.1 - 2 1301
BC079316.1 - 2 1276 7135022

View Full Product Details