Gene Transcripts

Gene Symbol
Adam28
Entrez Gene ID
290344
Gene Name
ADAM metallopeptidase domain 28
Gene Aliases
Chromosome Location
Chr. 15: 50358192 - 50425911 on Build Rnor_6.0
UniGene ID
Rn.97304
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181693.1 NP_859044.1 3 203
XM_008770820.1 XP_008769042.1 3 214
GenBank mRNA AY283187.1 - 3 203

View Full Product Details