Gene Transcripts

Gene Symbol
Yif1b
Entrez Gene ID
292768
Gene Name
Yip1 interacting factor homolog B (S. cerevisiae)
Gene Aliases
Yip1b
Chromosome Location
Chr. 1: 88182382 - 88192572 on Build Rnor_6.0
UniGene ID
Rn.154471
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014810.1 NP_001014810.1 2, 3 312
NM_198734.2 NP_942029.2 2, 3 322
XM_008759128.1 XP_008757350.1 2, 3 341
XM_008759129.1 XP_008757351.1 2, 3 321
GenBank mRNA BC058153.1 - 2, 3 315
CB576820.1 - 2, 3 265
CB797587.1 - 2, 3 322

Gene Symbol
LOC103689986
Entrez Gene ID
103689986
Gene Name
protein YIF1B
Gene Aliases
Chromosome Location
Chr. 1: 87180592 - 87190783 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228658.2 XP_006228720.1 2, 3 341
XM_006228660.2 XP_006228722.1 2, 3 321
XM_008759221.1 XP_008757443.1 2, 3 351
XM_008759222.1 XP_008757444.1 2, 3 370

View Full Product Details