Gene Transcripts

Gene Symbol
Tnmd
Entrez Gene ID
64104
Gene Name
tenomodulin
Gene Aliases
ChM1L
Chromosome Location
Chr. X: 104684676 - 104700173 on Build Rnor_6.0
UniGene ID
Rn.26691
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022290.1 NP_071626.1 6 825
GenBank mRNA AB055423.1 - 6 614
AF191769.1 - 6 825

View Full Product Details