Gene Transcripts

Gene Symbol
ESM1
Entrez Gene ID
11082
Gene Name
endothelial cell specific molecule 1
Gene Aliases
endocan
Chromosome Location
Chr. 5: 54977867 - 54985586 on Build GRCh38
UniGene ID
Hs.129944
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135604.1 NP_001129076.1 2 1584
NM_007036.4 NP_008967.1 3 1734
GenBank mRNA X89426.1 - 4 1692

View Full Product Details