Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022606.1 | NP_072128.1 | 6 | 557 | |
XM_008767296.1 | XP_008765518.1 | 5 | 2513 | ||
XM_008767297.1 | XP_008765519.1 | 4 | 2838 | ||
GenBank mRNA | AF095927.1 | - | 6 | 557 | |
BC062010.1 | - | 6 | 559 | 5621497 |