Gene Transcripts
- Gene Symbol
- Myo1c
- Entrez Gene ID
- 65261
- Gene Name
- myosin IC
- Gene Aliases
- Myr2
- Chromosome Location
- Chr. 10: 63803311 - 63819614 on Build Rnor_6.0
- UniGene ID
- Rn.55135
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023092.1 |
NP_075580.2
|
32 |
3632 |
|
GenBank mRNA |
X74800.1 |
- |
32 |
3632 |
|
View Full Product Details