Gene Transcripts

Gene Symbol
Psmc5
Entrez Gene ID
81827
Gene Name
proteasome (prosome, macropain) 26S subunit, ATPase, 5
Gene Aliases
Sug1
Chromosome Location
Chr. 10: 94445877 - 94451811 on Build Rnor_6.0
UniGene ID
Rn.10972
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031149.1 NP_112411.1 4, 5 287
GenBank mRNA AB000491.1 - 4, 5 287
BC058462.1 - 4, 5 283 6891005
D83521.1 - 4, 5 305
FQ213263.1 - 4, 5 304
FQ213735.1 - 4, 5 279
FQ233831.1 - 4, 5 290

View Full Product Details