Gene Transcripts
- Gene Symbol
- Hmga2
- Entrez Gene ID
- 84017
- Gene Name
- high mobility group AT-hook 2
- Gene Aliases
- Hmgic
- Chromosome Location
- Chr. 7: 65159944 - 65275408 on Build Rnor_6.0
- UniGene ID
- Rn.29981
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032070.1 |
NP_114459.1
|
5 |
773 |
|
GenBank mRNA |
U89695.1 |
- |
5 |
773 |
|
View Full Product Details