Gene Transcripts
- Gene Symbol
- Lmnb1
- Entrez Gene ID
- 116685
- Gene Name
- lamin B1
- Gene Aliases
- Chromosome Location
- Chr. 18: 51785111 - 51822264 on Build Rnor_6.0
- UniGene ID
- Rn.11362
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053905.1 |
NP_446357.1
|
2 |
826 |
|
GenBank mRNA |
U72353.1 |
- |
2 |
826 |
|
View Full Product Details