Gene Transcripts

Gene Symbol
Abtb2
Entrez Gene ID
171440
Gene Name
ankyrin repeat and BTB (POZ) domain containing 2
Gene Aliases
Cca3
Chromosome Location
Chr. 3: 93494706 - 93648750 on Build Rnor_6.0
UniGene ID
Rn.11149
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134403.2 NP_599230.2 1 1255
GenBank mRNA AB000216.1 - 1 1268

View Full Product Details