Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002261.2 | NP_002252.2 | 4 | 482 | |
NM_007333.2 | NP_031359.2 | 4 | 482 | ||
GenBank mRNA | AF078550.1 | - | 4 | 437 | |
AF350016.1 | - | 4 | 437 | ||
AF350017.1 | - | 4 | 437 | ||
AF461157.1 | - | 4 | 453 | ||
AK307860.1 | - | 4 | 281 | ||
AK311162.1 | - | 4 | 382 | ||
L14542.1 | - | 4 | 482 |