Gene Transcripts

Gene Symbol
Bambi
Entrez Gene ID
83837
Gene Name
BMP and activin membrane-bound inhibitor
Gene Aliases
Chromosome Location
Chr. 17: 56905419 - 56909675 on Build Rnor_6.0
UniGene ID
Rn.25267
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139082.3 NP_620782.1 3 439
GenBank mRNA AF387513.1 - 3 439
BC070516.1 - 3 683 7100920

View Full Product Details