Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008762466.1 | XP_008760688.1 | 6 | 994 | |
GenBank mRNA | AB029897.1 | - | 4 | 502 | |
AB180269.1 | - | 4 | 502 | ||
AB180270.1 | - | 4 | 502 | ||
AB180271.1 | - | 4 | 502 | ||
BC094306.1 | - | 4 | 490 | ||
CO405877.1 | - | 2 | 88 |