Gene Transcripts

Gene Symbol
Tomm20
Entrez Gene ID
266601
Gene Name
translocase of outer mitochondrial membrane 20 homolog (yeast)
Gene Aliases
Chromosome Location
Chr. 19: 59863421 - 59873411 on Build Rnor_6.0
UniGene ID
Rn.2143
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152935.1 NP_690918.1 5 773
GenBank mRNA BC078715.1 - 5 750
FQ224957.1 - 5 791
FQ230650.1 - 5 791
U21871.1 - 5 773

View Full Product Details