Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173125.1 | NP_775148.1 | 5 | 388 | |
XM_006253611.2 | XP_006253673.2 | 5 | 681 | ||
XM_008771487.1 | XP_008769709.1 | 5 | 681 | ||
GenBank mRNA | AF095585.1 | - | 5 | 388 | |
BC078693.1 | - | 5 | 380 | 7121395 |