Gene Transcripts

Gene Symbol
Phax
Entrez Gene ID
286917
Gene Name
phosphorylated adaptor for RNA export
Gene Aliases
Rnuxa
Chromosome Location
Chr. 18: 51662294 - 51678981 on Build Rnor_6.0
UniGene ID
Rn.10317
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173133.2 NP_775156.2 2 458
GenBank mRNA BC086410.1 - 2 428
BC097932.1 - 2 434 7386998
X67877.1 - 2 458

View Full Product Details