Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012478.1 | NP_001012496.1 | 5, 6 | 1549 | |
NM_007279.2 | NP_009210.1 | 5, 6 | 1549 | ||
XM_011526410.1 | XP_011524712.1 | 6, 7 | 1587 | ||
GenBank mRNA | AB451261.1 | - | 5, 6 | 494 | |
AB451387.1 | - | 5, 6 | 494 | ||
BC008740.2 | - | 5, 6 | 553 | 3346087 | |
BC030574.2 | - | 5, 6 | 1453 | 3687471 | |
X64044.1 | - | 5, 6 | 534 |