Gene Transcripts

Gene Symbol
RNF13
Entrez Gene ID
11342
Gene Name
ring finger protein 13
Gene Aliases
RZF
Chromosome Location
Chr. 3: 149812688 - 149962139 on Build GRCh38
UniGene ID
Hs.12333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007282.4 NP_009213.1 6 1135
NM_183381.2 NP_899237.1 5 1033
XM_005247092.3 XP_005247149.1 5 982
XM_011512373.2 XP_011510675.1 7 1140
XM_011512374.2 XP_011510676.1 6 1038
XM_011512376.2 XP_011510678.1 5 433
XM_017005654.1 XP_016861143.1 6 1214
XM_017005655.1 XP_016861144.1 5 1115
XM_017005656.1 XP_016861145.1 6 1084
XM_017005657.1 XP_016861146.1 6 1217
XM_017005658.1 XP_016861147.1 5 1115
XM_017005659.1 XP_016861148.1 4 852
XM_017005660.1 XP_016861149.1 4 852
GenBank mRNA AF037204.1 - 5 502
AF070558.1 - 5 493
AK090638.1 - 5 1013
AK090771.1 - 2 112
AK313304.1 - 5 517
BC009781.1 - 5 510 3683407
BC009803.1 - 6 613 4274992
BC017878.1 - 5 475
BG546092.1 - 3 296
BG708380.1 - 5 505
BM556246.1 - 4 378
CR456804.1 - 4 351

View Full Product Details