Gene Transcripts

Gene Symbol
MTF2
Entrez Gene ID
22823
Gene Name
metal response element binding transcription factor 2
Gene Aliases
M96, PCL2, TDRD19A, dJ976O13.2
Chromosome Location
Chr. 1: 93079219 - 93139081 on Build GRCh38
UniGene ID
Hs.31016
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164391.1 NP_001157863.1 3 370
NM_001164392.1 NP_001157864.1 2 450
NM_007358.3 NP_031384.1 2 450
XM_011541016.1 XP_011539318.1 2 402
XM_011541017.2 XP_011539319.1 2 466
XM_011541019.1 XP_011539321.1 3 322
XM_017000676.1 XP_016856165.1 2 305
XM_017000677.1 XP_016856166.1 2 402
XM_017000678.1 XP_016856167.1 3 359
XM_017000679.1 XP_016856168.1 2 402
GenBank mRNA AF072814.1 - 1 144
AJ010014.1 - 2 401
AK290318.1 - 2 402
AK302776.1 - 3 309
BC001897.1 - 2 369
BC010013.2 - 2 374 4308565
DB173315.1 - 2 450

View Full Product Details