Gene Transcripts
- Gene Symbol
- Ntsr2
- Entrez Gene ID
- 64636
- Gene Name
- neurotensin receptor 2
- Gene Aliases
- Chromosome Location
- Chr. 6: 41917094 - 41923780 on Build Rnor_6.0
- UniGene ID
- Rn.127792
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022695.2 |
NP_073186.1
|
4 |
1399 |
|
GenBank mRNA |
X97121.1 |
- |
4 |
1401 |
|
View Full Product Details