Gene Transcripts

Gene Symbol
Atic
Entrez Gene ID
81643
Gene Name
5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase
Gene Aliases
Chromosome Location
Chr. 9: 78862013 - 78882061 on Build Rnor_6.0
UniGene ID
Rn.15114
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031014.2 NP_112276.2 5, 6 436
GenBank mRNA BC072496.1 - 5, 6 436
D89514.1 - 5, 6 429

View Full Product Details