Gene Transcripts
- Gene Symbol
- Trim3
- Entrez Gene ID
- 83616
- Gene Name
- tripartite motif-containing 3
- Gene Aliases
- Berp, Rnf22
- Chromosome Location
- Chr. 1: 170441662 - 170464341 on Build Rnor_6.0
- UniGene ID
- Rn.203982
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031786.1 |
NP_113974.1
|
3, 4 |
579 |
|
GenBank mRNA |
AF036255.1 |
- |
3, 4 |
579 |
|
View Full Product Details