Gene Transcripts

Gene Symbol
Ghsr
Entrez Gene ID
84022
Gene Name
growth hormone secretagogue receptor
Gene Aliases
Chromosome Location
Chr. 2: 113066885 - 113070261 on Build Rnor_6.0
UniGene ID
Rn.74241
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032075.3 NP_114464.1 1 188
GenBank mRNA AB001982.1 - 1 188
U94321.1 - 1 188

View Full Product Details