Gene Transcripts
- Gene Symbol
- CCNDBP1
- Entrez Gene ID
- 23582
- Gene Name
- cyclin D1 binding protein 1
- Gene Aliases
- DIP1, GCIP, HHM
- Chromosome Location
- Chr. 15: 43185118 - 43197177 on Build GRCh38
- UniGene ID
- Hs.36794
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012142.4 |
NP_036274.3
|
3, 4 |
488 |
|
NR_027514.2 |
|
3, 4 |
488 |
|
GenBank mRNA |
AF082569.1 |
- |
3, 4 |
347 |
|
AF087852.1 |
- |
3, 4 |
333 |
|
AF113535.1 |
- |
1, 2 |
297 |
|
AF132034.1 |
- |
3, 4 |
303 |
|
AF246144.1 |
- |
3, 4 |
305 |
|
AK075146.1 |
- |
3, 4 |
317 |
|
AK075296.1 |
- |
1, 2 |
302 |
|
AK225143.1 |
- |
3, 4 |
303 |
|
AK225645.1 |
- |
3, 4 |
317 |
|
AK290665.1 |
- |
3, 4 |
317 |
|
AK295017.1 |
- |
3, 4 |
317 |
|
AK308817.1 |
- |
2, 3 |
912 |
|
AL552334.3 |
- |
3, 4 |
427 |
|
BC009689.1 |
- |
3, 4 |
331 |
3896421 |
CR450331.1 |
- |
3, 4 |
257 |
|
DA506028.1 |
- |
3, 4 |
477 |
|
View Full Product Details