Gene Transcripts

Gene Symbol
Mfn2
Entrez Gene ID
64476
Gene Name
mitofusin 2
Gene Aliases
HSG
Chromosome Location
Chr. 5: 164684244 - 164715412 on Build Rnor_6.0
UniGene ID
Rn.8570
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130894.4 NP_570964.4 2 245
XM_006239402.2 XP_006239464.1 3 398
XM_006239403.1 XP_006239465.1 3 427
XM_008764288.1 XP_008762510.1 4 415
GenBank mRNA AB084165.1 - 1 98
BC070880.1 - 2 249 7096620
U41803.2 - 3 263

Gene Symbol
LOC100911485
Entrez Gene ID
100911485
Gene Name
mitofusin-2-like
Gene Aliases
Chromosome Location
Chr. 5: 164560437 - 164586254 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750086.3 XP_003750134.1 718

View Full Product Details