Gene Transcripts
- Gene Symbol
- FBXO25
- Entrez Gene ID
- 26260
- Gene Name
- F-box protein 25
- Gene Aliases
- FBX25
- Chromosome Location
- Chr. 8: 406808 - 477967 on Build GRCh38
- UniGene ID
- Hs.438454
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012173.3 |
NP_036305.2
|
3, 4 |
514 |
|
NM_183420.1 |
NP_904356.1
|
4, 5 |
564 |
|
NM_183421.1 |
NP_904357.1
|
4, 5 |
564 |
|
XM_011534748.2 |
XP_011533050.1
|
4, 5 |
337 |
|
XM_011534749.1 |
XP_011533051.1
|
3, 4 |
274 |
|
XM_017013313.1 |
XP_016868802.1
|
4, 5 |
388 |
|
GenBank mRNA |
AB070270.1 |
- |
3, 4 |
298 |
|
AF419858.1 |
- |
4, 5 |
564 |
|
AK302334.1 |
- |
4, 5 |
323 |
|
BC020249.2 |
- |
2, 3 |
256 |
4652811 |
BC050393.1 |
- |
4, 5 |
417 |
5769518 |
- Gene Symbol
- LOC107985330
- Entrez Gene ID
- 107985330
- Gene Name
- F-box only protein 25-like
- Gene Aliases
- Chromosome Location
- Chr. 1: 224027677 - 224035390 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC728323
- Entrez Gene ID
- 728323
- Gene Name
- uncharacterized LOC728323
- Gene Aliases
- Chromosome Location
- Chr. 2: 242088633 - 242160503 on Build GRCh38
- UniGene ID
- Hs.380737
- Species
- Homo sapiens
View Full Product Details