Gene Transcripts
- Gene Symbol
- GCA
- Entrez Gene ID
- 25801
- Gene Name
- grancalcin
- Gene Aliases
- GCL
- Chromosome Location
- Chr. 2: 162318768 - 162371605 on Build GRCh38
- UniGene ID
- Hs.377894
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012198.3 |
NP_036330.1
|
2, 3 |
376 |
|
XM_005246446.2 |
XP_005246503.1
|
2, 3 |
317 |
|
XM_006712398.3 |
XP_006712461.1
|
2, 3 |
549 |
|
XM_006712400.3 |
XP_006712463.1
|
3, 4 |
627 |
|
XM_006712401.3 |
XP_006712464.1
|
2, 3 |
628 |
|
XM_011510926.2 |
XP_011509228.1
|
2, 3 |
328 |
|
XM_011510927.2 |
XP_011509229.1
|
2, 3 |
323 |
|
XM_017003764.1 |
XP_016859253.1
|
3, 4 |
640 |
|
XM_017003765.1 |
XP_016859254.1
|
3, 4 |
726 |
|
XM_017003766.1 |
XP_016859255.1
|
3, 4 |
523 |
|
XM_017003767.1 |
XP_016859256.1
|
2, 3 |
399 |
|
XM_017003768.1 |
XP_016859257.1
|
2, 3 |
394 |
|
XM_017003769.1 |
XP_016859258.1
|
2, 3 |
549 |
|
GenBank mRNA |
AB209768.1 |
- |
2, 3 |
265 |
|
AK295413.1 |
- |
2, 3 |
309 |
|
AK312349.1 |
- |
2, 3 |
325 |
|
BC005214.1 |
- |
2, 3 |
299 |
3996889 |
M81637.1 |
- |
2, 3 |
319 |
|
View Full Product Details