Gene Transcripts

Gene Symbol
Plod3
Entrez Gene ID
288583
Gene Name
procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3
Gene Aliases
Chromosome Location
Chr. 12: 22716421 - 22726982 on Build Rnor_6.0
UniGene ID
Rn.90152
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178101.3 NP_835202.2 5 866
GenBank mRNA AJ430859.1 - 5 605
BC085683.1 - 5 646 7110561

View Full Product Details