Gene Transcripts

Gene Symbol
Egln1
Entrez Gene ID
308913
Gene Name
egl-9 family hypoxia-inducible factor 1
Gene Aliases
HIF-PH2, HPH-2, PHD-2, PHD2
Chromosome Location
Chr. 19: 57660194 - 57701158 on Build Rnor_6.0
UniGene ID
Rn.232203
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772679.1 XP_008770901.1 3 1478
GenBank mRNA AY228140.1 - 1 263
BC081694.1 - 1 507

Gene Symbol
LOC690182
Entrez Gene ID
690182
Gene Name
similar to Egl nine homolog 1 (Hypoxia-inducible factor prolyl hydroxylase 2) (HIF-prolyl hydroxylase 2) (HIF-PH2) (HPH-2) (SM-20)
Gene Aliases
Chromosome Location
Chr. 1: 173283884 - 173291061 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759848.1 XP_008758070.1 1 503

View Full Product Details