Gene Transcripts

Gene Symbol
Ncr3
Entrez Gene ID
294251
Gene Name
natural cytotoxicity triggering receptor 3
Gene Aliases
1C7, Nkp30
Chromosome Location
Chr. 20: 5170166 - 5175773 on Build Rnor_6.0
UniGene ID
Rn.203206
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181822.2 NP_861543.2 3 681
XM_006256054.2 XP_006256116.1 3 851
GenBank mRNA AY273824.1 - 3 802

View Full Product Details