Gene Transcripts

Gene Symbol
Atp6v1e1
Entrez Gene ID
297566
Gene Name
ATPase, H+ transporting, lysosomal V1 subunit E1
Gene Aliases
Atp6e, Atp6e2, P31, Vma4
Chromosome Location
Chr. 4: 153351434 - 153373558 on Build Rnor_6.0
UniGene ID
Rn.103171
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198745.2 NP_942040.1 3 255
GenBank mRNA BC059155.1 - 3 259
FQ212928.1 - 3 255
FQ212966.1 - 3 250
FQ213001.1 - 3 255
FQ213103.1 - 3 252
FQ213345.1 - 3 257
FQ213479.1 - 3 256
FQ213531.1 - 3 257
FQ213667.1 - 3 273
FQ213669.1 - 3 257
FQ213903.1 - 3 251
FQ214240.1 - 3 251
FQ214254.1 - 3 257
FQ214381.1 - 3 255
FQ214406.1 - 3 250
FQ214476.1 - 3 260
FQ214497.1 - 3 223
FQ214920.1 - 3 255
FQ216629.1 - 3 255
FQ224865.1 - 3 256
FQ226034.1 - 3 250
FQ230006.1 - 3 255
FQ230733.1 - 3 250
FQ233079.1 - 3 255
FQ233569.1 - 3 257

View Full Product Details