Gene Transcripts

Gene Symbol
Slc35b1
Entrez Gene ID
287642
Gene Name
solute carrier family 35, member B1
Gene Aliases
UGTrel1
Chromosome Location
Chr. 10: 83201532 - 83208605 on Build Rnor_6.0
UniGene ID
Rn.124916
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199081.1 NP_954512.1 7 804
GenBank mRNA AY349135.1 - 7 804
BC085347.1 - 7 839 7305181
D87991.1 - 7 797
FQ233464.1 - 7 827

View Full Product Details