Gene Transcripts

Gene Symbol
Nudt22
Entrez Gene ID
293703
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 22
Gene Aliases
Chromosome Location
Chr. 1: 222247500 - 222250983 on Build Rnor_6.0
UniGene ID
Rn.1934
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199090.2 NP_954521.1 2 284
XM_006230760.2 XP_006230822.1 1 186
GenBank mRNA BC060593.1 - 2 285
FQ212766.1 - 2 231
FQ215165.1 - 2 212
FQ219356.1 - 2 235

View Full Product Details