Gene Transcripts

Gene Symbol
Plekha4
Entrez Gene ID
308584
Gene Name
pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 4
Gene Aliases
Chromosome Location
Chr. 1: 101517740 - 101540802 on Build Rnor_6.0
UniGene ID
Rn.99114
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199101.1 NP_954532.1 8 1051
XM_006229071.1 XP_006229133.1 7 991
XM_006229072.1 XP_006229134.1 8 1207
GenBank mRNA BC061738.1 - 8 1051

View Full Product Details