Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012137.3 | NP_036269.1 | 1, 2 | 469 | |
GenBank mRNA | AK312432.1 | - | 1, 2 | 430 | |
BC033680.1 | - | 1, 2 | 407 | 5189970 | |
BC043235.2 | - | 1, 2 | 191 | ||
DC324361.1 | - | 1, 2 | 471 | ||
EU794605.1 | - | 1, 2 | 407 |