Gene Transcripts
- Gene Symbol
- DNAI1
- Entrez Gene ID
- 27019
- Gene Name
- dynein axonemal intermediate chain 1
- Gene Aliases
- CILD1, DIC1, ICS1, PCD
- Chromosome Location
- Chr. 9: 34458752 - 34520989 on Build GRCh38
- UniGene ID
- Hs.112667
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281428.1 |
NP_001268357.1
|
2, 3 |
336 |
|
NM_012144.3 |
NP_036276.1
|
2, 3 |
336 |
|
XM_006716758.3 |
XP_006716821.1
|
1, 2 |
34 |
|
XM_011517846.1 |
XP_011516148.1
|
2, 3 |
336 |
|
XM_011517847.2 |
XP_011516149.1
|
2, 3 |
336 |
|
XM_011517848.1 |
XP_011516150.1
|
2, 3 |
336 |
|
XM_011517849.1 |
XP_011516151.1
|
2, 3 |
336 |
|
XM_011517850.2 |
XP_011516152.1
|
2, 3 |
336 |
|
XM_017014625.1 |
XP_016870114.1
|
2, 3 |
336 |
|
GenBank mRNA |
AF091619.1 |
- |
2, 3 |
253 |
|
AK290696.1 |
- |
2, 3 |
260 |
|
AK302499.1 |
- |
2, 3 |
199 |
|
AK303937.1 |
- |
2, 3 |
225 |
|
AK314639.1 |
- |
2, 3 |
248 |
|
BC030583.2 |
- |
2, 3 |
264 |
4837991 |
BM013886.1 |
- |
2, 3 |
275 |
|
DB033871.1 |
- |
2, 3 |
260 |
|
DB046118.1 |
- |
2, 3 |
337 |
|
View Full Product Details