Gene Transcripts

Gene Symbol
Ephx1
Entrez Gene ID
25315
Gene Name
epoxide hydrolase 1, microsomal (xenobiotic)
Gene Aliases
MEH8
Chromosome Location
Chr. 13: 99271390 - 99300580 on Build Rnor_6.0
UniGene ID
Rn.3603
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034090.3 NP_001029262.1 2 223
NM_012844.3 NP_036976.2 2 252
GenBank mRNA BC061568.1 - 2 183
M26125.1 - 228

View Full Product Details