Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012880.1 | NP_037012.1 | 2 | 303 | |
XM_006251030.2 | XP_006251092.1 | 2 | 349 | ||
GenBank mRNA | BC061861.1 | - | 2 | 254 | |
X68041.1 | - | 2 | 278 | ||
X94371.1 | - | 2 | 271 | ||
Z24721.1 | - | 2 | 303 |