Gene Transcripts

Gene Symbol
Dlx5
Entrez Gene ID
25431
Gene Name
distal-less homeobox 5
Gene Aliases
RDLX
Chromosome Location
Chr. 4: 32387741 - 32392007 on Build Rnor_6.0
UniGene ID
Rn.11248
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012943.1 NP_037075.1 2 586
GenBank mRNA AB073716.1 - 2 308
D31734.1 - 2 583
L24443.1 - 2 586

View Full Product Details