Gene Transcripts

Gene Symbol
Slc18a2
Entrez Gene ID
25549
Gene Name
solute carrier family 18 (vesicular monoamine transporter), member 2
Gene Aliases
MNAT, VMAT-2, VMAT2
Chromosome Location
Chr. 1: 280422237 - 280457964 on Build Rnor_6.0
UniGene ID
Rn.9686
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013031.1 NP_037163.1 3 264
XM_006231664.2 XP_006231726.1 3 1105
XM_008760515.1 XP_008758737.1 2 464
XM_008760516.1 XP_008758738.1 3 1110
XM_008760517.1 XP_008758739.1 3 1105
XM_008760518.1 XP_008758740.1 2 464
XM_008760519.1 XP_008758741.1 2 464
XM_008760520.1 XP_008758742.1 2 464
GenBank mRNA L00603.1 - 3 264
M97381.1 - 3 235

View Full Product Details