Gene Transcripts

Gene Symbol
Pkn1
Entrez Gene ID
29355
Gene Name
protein kinase N1
Gene Aliases
PAK-1, Prkcl1
Chromosome Location
Chr. 19: 24803524 - 24831500 on Build Rnor_6.0
UniGene ID
Rn.49880
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017175.2 NP_058871.2 5 851
XM_006255245.2 XP_006255307.1 5 1891
GenBank mRNA BC061836.1 - 5 752
D26180.1 - 5 802
FQ222217.1 - 5 851
L35634.1 - 5 694

View Full Product Details