Gene Transcripts

Gene Symbol
Htr2a
Entrez Gene ID
29595
Gene Name
5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled
Gene Aliases
5-HT2A, 5Ht-2
Chromosome Location
Chr. 15: 56666152 - 56732469 on Build Rnor_6.0
UniGene ID
Rn.10294
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017254.1 NP_058950.1 1 191
GenBank mRNA M30705.1 - 1 191
M64867.1 - 597
X13971.1 - 734

View Full Product Details