Gene Transcripts

Gene Symbol
Slc23a1
Entrez Gene ID
50621
Gene Name
solute carrier family 23 (ascorbic acid transporter), member 1
Gene Aliases
SVCT1
Chromosome Location
Chr. 18: 28414008 - 28428151 on Build Rnor_6.0
UniGene ID
Rn.144579
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017315.2 NP_059011.1 2 152
XM_006254602.2 XP_006254664.1 4 533
GenBank mRNA AF080452.1 - 2 152
BC078851.1 - 2 152 7127602

View Full Product Details