Gene Transcripts
- Gene Symbol
- Chrng
- Entrez Gene ID
- 25753
- Gene Name
- cholinergic receptor, nicotinic, gamma (muscle)
- Gene Aliases
- ACHRG
- Chromosome Location
- Chr. 9: 94302218 - 94308591 on Build Rnor_6.0
- UniGene ID
- Rn.44634
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019145.1 |
NP_062018.1
|
2 |
301 |
|
GenBank mRNA |
X74834.1 |
- |
2 |
301 |
|
View Full Product Details