Gene Transcripts
- Gene Symbol
- Fgf18
- Entrez Gene ID
- 29369
- Gene Name
- fibroblast growth factor 18
- Gene Aliases
- Chromosome Location
- Chr. 10: 18047109 - 18082290 on Build Rnor_6.0
- UniGene ID
- Rn.44478
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019199.1 |
NP_062072.1
|
2, 3 |
365 |
|
GenBank mRNA |
AB004638.1 |
- |
2, 3 |
365 |
|
View Full Product Details