Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020100.2 | NP_064485.2 | 3 | 679 | |
GenBank mRNA | AB042889.1 | - | 3 | 677 | |
AF181552.2 | - | 3 | 679 | ||
FQ214551.1 | - | 3 | 673 | ||
FQ220330.1 | - | 3 | 660 | ||
FQ220789.1 | - | 3 | 681 |